missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)piperidine, 97%

Catalog No. AAH2695703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2695703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2695703 Supplier Thermo Scientific Chemicals Supplier No. H2695703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 216854-23-8 | C10H20N2O2 | 200.282 g/mol

(S)-3-(Boc-amino)piperidine, is used as an intermediate for pigments. It is also used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also an useful intermediate for the synthesis of a variety of chiral aminopiperidinyl quinolones as potent antibacterial agents against resistant pathogens; and alkynylpyrimidine amide derivatives as orally available inhibitors of Tie-2 kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-(Boc-amino)piperidine, is used as an intermediate for pigments. It is also used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also an useful intermedia

Chemical Identifiers

CAS 216854-23-8
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093383
InChI Key WUOQXNWMYLFAHT-QMMMGPOBSA-N
Synonym s-3-boc-aminopiperidine, s-3-n-boc-aminopiperidine, s-tert-butyl piperidin-3-ylcarbamate, s-3-boc-amino piperidine, tert-butyl n-3s-piperidin-3-yl carbamate, s-3-tert-butoxycarbonylamino piperidine, tert-butyl s-piperidin-3-ylcarbamate, s-3-n-boc-amino piperidine, s---3-tert-butoxycarbonylaminopiperidine, 3s-3-aminopiperidine, 3-boc protected
PubChem CID 1514171
IUPAC Name tert-butyl N-[(3S)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Melting Point 122°C to 127°C
Solubility Information Freely soluble in ethanol.
Quantity 1 g
Optical Rotation −15.0° (c=1 in MeOH)
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-3-(Boc-amino)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only