missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%

Catalog No. AAH5217303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5217303 1 g
AAH52173MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5217303 Supplier Thermo Scientific Chemicals Supplier No. H5217303

CAS: 132549-43-0 | C12H23NO4 | 245.32 g/mol

(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of carba peptides and building block for β-peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of ″carba peptides″ and building block for β-peptides.

Solubility
Very slightly soluble (1.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 132549-43-0
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.32
MDL Number MFCD02101665
InChI Key XRVAMBSTOWHUMM-VIFPVBQESA-N
Synonym boc-l-beta-homoleucine, s-3-tert-butoxycarbonyl amino-5-methylhexanoic acid, boc-beta-homoleu-oh, boc-l-beta-hleu-oh, hexanoic acid, 3-1,1-dimethylethoxy carbonyl amino-5-methyl-, 3s, 3s-3-tert-butoxycarbonyl amino-5-methylhexanoic acid, ambotzbaa6190, boc-leu-c*ch2 oh, pubchem12105, rarechem em wb 0131
PubChem CID 2761526
IUPAC Name (3S)-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)C[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 53°C to 55°C
Quantity 1 g
Beilstein 4251252
Solubility Information Very slightly soluble (1.1g/L at 25°C).
Formula Weight 245.32
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-5-methylhexanoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only