missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%

Catalog No. AAH52174MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52174MD 250 mg
AAH5217403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52174MD Supplier Thermo Scientific Chemicals Supplier No. H52174MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51871-62-6 | C15H21NO4 | 279.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51871-62-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD01076271
InChI Key ACKWQHCPHJQANL-LBPRGKRZSA-N
Synonym s-3-boc-amino-4-phenylbutyric acid, boc-l-beta-homophenylalanine, boc-beta-hophe-oh, 3s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, 3r-3-tert-butoxy carbonyl amino-4-phenylbutanoic acid, boc-hophe-oh, pubchem12107, tmba023, 3s-n-boc-3-benzyl-beta-alanine
PubChem CID 2761538
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1

Specifications

Melting Point 101°C to 107°C
Beilstein 3060457
Quantity 250 mg
Optical Rotation −19.0° (c=1 in Dichloromethane)
Formula Weight 279.34
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-4-phenylbutyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only