missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%

CAS: 51871-62-6 | C15H21NO4 | 279.34 g/mol

Supplier:  Thermo Scientific Chemicals H5217403

Quantity 1 g

 View more versions of this product

Catalog No. AAH5217403


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51871-62-6
279.34
ACKWQHCPHJQANL-LBPRGKRZSA-N
2761538
CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1
C15H21NO4
MFCD01076271
s-3-boc-amino-4-phenylbutyric acid, boc-l-beta-homophenylalanine, boc-beta-hophe-oh, 3s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, 3r-3-tert-butoxy carbonyl amino-4-phenylbutanoic acid, boc-hophe-oh, pubchem12107, tmba023, 3s-n-boc-3-benzyl-beta-alanine
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid

Specifications

101°C to 107°C
51871-62-6
MFCD01076271
s-3-boc-amino-4-phenylbutyric acid, boc-l-beta-homophenylalanine, boc-beta-hophe-oh, 3s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, 3r-3-tert-butoxy carbonyl amino-4-phenylbutanoic acid, boc-hophe-oh, pubchem12107, tmba023, 3s-n-boc-3-benzyl-beta-alanine
CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1
1 g
2761538
279.34
(S)-3-(Boc-amino)-4-phenylbutyric acid
C15H21NO4
3060457
ACKWQHCPHJQANL-LBPRGKRZSA-N
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
279.34
−19.0° (c=1 in Dichloromethane)
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions