missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%

Catalog No. p-7056870
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66034MD 250 mg
AAH6603403 1 g
AAH6603406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66034MD Supplier Thermo Scientific Chemicals Supplier No. H66034MD
Only null left

CAS: 204512-95-8 | C4H10ClNO | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 204512-95-8
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD08445642
InChI Key MHOVLDXJDIEEMJ-WCCKRBBISA-N
Synonym s-tetrahydrofuran-3-amine hydrochloride, s-3-aminotetrahydrofuran hydrochloride, 3s-oxolan-3-amine hydrochloride, s-3-aminotetrahydrofuran hcl, s-tetrahydrofuran-3-amine hcl, s-tetrahydro-furan-3-ylamine hydrochloride, 3-furanamine, tetrahydro-, hydrochloride, 3s, s-3-aminotetrahydrofuranhydrochloride, pubchem15126, ksc916a5t
PubChem CID 18664284
IUPAC Name (3S)-oxolan-3-amine;hydrochloride
SMILES C1COCC1N.Cl

Specifications

Quantity 250 mg
Beilstein 14637084
Formula Weight 123.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (S)-(+)-3-Aminotetrahydrofuran hydrochloride
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only