missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Aminopiperidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH2770003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2770003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2770003 Supplier Thermo Scientific Chemicals Supplier No. H2770003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334618-07-4
Molecular Formula C5H14Cl2N2
Molecular Weight (g/mol) 173.08
MDL Number MFCD03427036
InChI Key GGPNYXIOFZLNKW-UHFFFAOYNA-N
Synonym s-3-aminopiperidine dihydrochloride, s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine dihydrochloride, s-3-aminopiperidine 2hcl, 3s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine 2hcl, 3-piperidinamine, dihydrochloride, 3s, 3s-3-piperidinamine dihydrochloride, 3s-3-aminopiperidine dihydrochloride, s-piperidin-3-ylamine dihydrochloride
PubChem CID 16218278
IUPAC Name (3S)-piperidin-3-amine;dihydrochloride
SMILES Cl.Cl.NC1CCCNC1

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Optical Rotation +1.6° (c=1 in Water)
Formula Weight 173.08
Percent Purity 98%
Chemical Name or Material (S)-(+)-3-Aminopiperidine dihydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only