missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee

Catalog No. p-4510672
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC362040010 1 g
AC362040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362040010 Supplier Thermo Scientific Chemicals Supplier No. 362040010

CAS: 167834-24-4 | C11H16NO2 | 194.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167834-24-4
Molecular Formula C11H16NO2
Molecular Weight (g/mol) 194.25
MDL Number MFCD02259715
InChI Key NUWRDXMXYDWUAN-JTQLQIEISA-O
Synonym s-ethyl 3-amino-3-phenylpropanoate hydrochloride, s-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, ethyl 3s-3-amino-3-phenylpropanoate hydrochloride, 3s-3-amino-3-phenylpropanoic acid ethyl ester hcl, s-beta-phenylalanine ethyl ester hydrochloride, pubchem13852, s-b-phenylalanine ethyl ester hcl, ethyl s-beta-aminobenzenepropanoate hcl, s-3-amino-3-phenylpropanoicacidethylesterhcl, ethyl 3s-3-amino-3-phenylpropanoate, chloride
PubChem CID 22831526
IUPAC Name ethyl (3S)-3-amino-3-phenylpropanoate;hydrochloride
SMILES CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1

Specifications

Melting Point 117.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 229.71
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Hazard Category Warning

RUO – Research Use Only