missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals

Catalog No. p-57683
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362170010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC362170010 Supplier Thermo Scientific Chemicals Supplier No. 362170010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82769-76-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD01311768
InChI Key SEQXIQNPMQTBGN-VIFPVBQESA-N
Synonym s-3-amino-3-phenylpropan-1-ol, 3s-3-amino-3-phenylpropan-1-ol, s-1-phenyl-3-propanolamine, s-beta-phenylalaninol, s-3-phenyl-beta-alaninol, s-3-amino-3-phenyl-1-propanol, s-3-amino-3-phenyl-propan-1-ol, 3s-3-amino-3-phenyl-propan-1-ol, pubchem13854, ksc491q5n
PubChem CID 2734520
IUPAC Name (3S)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless or White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.21
Percent Purity 95%,98% ee
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (S)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only