missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-3-Amino-2-piperidone 98.0+%, TCI America™
$469.75 - $1422.27
Chemical Identifiers
| CAS | 34294-79-6 |
|---|---|
| Molecular Formula | C5H10N2O |
| Molecular Weight (g/mol) | 114.15 |
| MDL Number | MFCD09258624 |
| InChI Key | YCCMTCQQDULIFE-UHFFFAOYNA-N |
| PubChem CID | 7200369 |
| IUPAC Name | 3-aminopiperidin-2-one |
| SMILES | NC1CCCNC1=O |
Chemical Identifiers
| 34294-79-6 | |
| 114.15 | |
| YCCMTCQQDULIFE-UHFFFAOYNA-N | |
| 3-aminopiperidin-2-one |
| C5H10N2O | |
| MFCD09258624 | |
| 7200369 | |
| NC1CCCNC1=O |
Specifications
| 34294-79-6 | |
| Yellow | |
| MFCD09258624 | |
| YCCMTCQQDULIFE-UHFFFAOYNA-N | |
| 3-aminopiperidin-2-one | |
| 7200369 | |
| ≥98.0% (GC) | |
| (S)-(-)-3-Amino-2-piperidone |
| 87°C | |
| C5H10N2O | |
| 1 g | |
| NC1CCCNC1=O | |
| 114.15 | |
| 114.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)