missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Amino-1-carbobenzoxypyrrolidine Hydrochloride 98.0+%, TCI America™
$288.77 - $1110.82
Chemical Identifiers
| CAS | 550378-39-7 |
|---|---|
| Molecular Formula | C12H17ClN2O2 |
| Molecular Weight (g/mol) | 256.73 |
| MDL Number | MFCD06796559 |
| InChI Key | QNQVBYGRFHOBNO-MERQFXBCSA-N |
| Synonym | (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride |
| PubChem CID | 20820180 |
| IUPAC Name | benzyl (3S)-3-aminopyrrolidine-1-carboxylate;hydrochloride |
| SMILES | C1CN(CC1N)C(=O)OCC2=CC=CC=C2.Cl |
Chemical Identifiers
| 550378-39-7 | |
| 256.73 | |
| QNQVBYGRFHOBNO-MERQFXBCSA-N | |
| 20820180 | |
| C1CN(CC1N)C(=O)OCC2=CC=CC=C2.Cl |
| C12H17ClN2O2 | |
| MFCD06796559 | |
| (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride | |
| benzyl (3S)-3-aminopyrrolidine-1-carboxylate;hydrochloride |
Specifications
| 550378-39-7 | |
| C12H17ClN2O2 | |
| 1 g | |
| QNQVBYGRFHOBNO-MERQFXBCSA-N | |
| benzyl (3S)-3-aminopyrrolidine-1-carboxylate;hydrochloride | |
| 20820180 | |
| ≥98.0% (HPLC,N) | |
| (S)-3-Amino-1-carbobenzoxypyrrolidine Hydrochloride |
| White-Yellow | |
| MFCD06796559 | |
| (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride | |
| C1CN(CC1N)C(=O)OCC2=CC=CC=C2.Cl | |
| 256.73 | |
| 256.73 | |
| Crystalline Powder |
Safety and Handling
TSCA : No