missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Amino-1,2-propanediol 98.0+%, TCI America™
$65.46 - $202.15
Chemical Identifiers
| CAS | 61278-21-5 |
|---|---|
| Molecular Formula | C3H9NO2 |
| Molecular Weight (g/mol) | 91.11 |
| MDL Number | MFCD00798260 |
| InChI Key | KQIGMPWTAHJUMN-VKHMYHEASA-N |
| Synonym | s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol |
| PubChem CID | 440902 |
| ChEBI | CHEBI:1456 |
| IUPAC Name | (2S)-3-aminopropane-1,2-diol |
| SMILES | C(C(CO)O)N |
Chemical Identifiers
| 61278-21-5 | |
| 91.11 | |
| KQIGMPWTAHJUMN-VKHMYHEASA-N | |
| 440902 | |
| (2S)-3-aminopropane-1,2-diol |
| C3H9NO2 | |
| MFCD00798260 | |
| s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol | |
| CHEBI:1456 | |
| C(C(CO)O)N |
Specifications
| 61278-21-5 | |
| White-Yellow | |
| C3H9NO2 | |
| 1 g | |
| s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol | |
| C(C(CO)O)N | |
| 91.11 | |
| CHEBI:1456 | |
| ≥98.0% (GC,T) | |
| (S)-3-Amino-1,2-propanediol |
| 55°C | |
| 130°C | |
| MFCD00798260 | |
| 3259 | |
| KQIGMPWTAHJUMN-VKHMYHEASA-N | |
| (2S)-3-aminopropane-1,2-diol | |
| 440902 | |
| 91.11 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-3504
RTECSNumber : TY2800000
TSCA : No