missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Amino-1,2-propanediol, 98%

Catalog No. AAH2674306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2674306 5 g
AAH2674303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2674306 Supplier Thermo Scientific Chemicals Supplier No. H2674306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61278-21-5 | C3H9NO2 | 91.11 g/mol

(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 61278-21-5
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00798260
InChI Key KQIGMPWTAHJUMN-VKHMYHEASA-N
Synonym s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
PubChem CID 440902
ChEBI CHEBI:1456
IUPAC Name (2S)-3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Melting Point 54°C to 56°C
Density 1.175
Boiling Point 117°C to 119°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 5 g
UN Number UN3259
Beilstein 1719122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −28° (c=4, 5M HCl)
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material (S)-(-)-3-Amino-1,2-propanediol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only