missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%
CAS: 33878-99-8 | C11H14O2 | 178.231 g/mol
$233.01 - $727.30
Chemical Identifiers
| CAS | 33878-99-8 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.231 |
| MDL Number | MFCD00213797 |
| InChI Key | DNHDRUMZDHWHKG-NSHDSACASA-N |
| Synonym | s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # |
| PubChem CID | 736069 |
| IUPAC Name | (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
| SMILES | CC12CCC(=O)C=C1CCCC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL13334ME
|
Thermo Scientific Chemicals
L13334ME |
500 mg |
Each for $233.01
|
|
|||||
|
AAL1333404
|
Thermo Scientific Chemicals
L1333404 |
2 g |
Each for $727.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 33878-99-8 | |
| 178.231 | |
| DNHDRUMZDHWHKG-NSHDSACASA-N | |
| 736069 | |
| CC12CCC(=O)C=C1CCCC2=O |
| C11H14O2 | |
| MFCD00213797 | |
| s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # | |
| (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
Specifications
| 33878-99-8 | |
| C11H14O2 | |
| 500 mg | |
| s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # | |
| +100° (c=1 in Benzene) | |
| (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione | |
| 736069 | |
| 99% |
| 48°C to 52°C | |
| MFCD00213797 | |
| 1569979 | |
| DNHDRUMZDHWHKG-NSHDSACASA-N | |
| CC12CCC(=O)C=C1CCCC2=O | |
| 178.231 | |
| 178.23 | |
| (S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only