missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-(tert-Butoxycarbonylamino)butyric Acid 98.0+%, TCI America™
$76.43 - $190.13
Chemical Identifiers
| CAS | 34306-42-8 |
|---|---|
| Molecular Formula | C9H17NO4 |
| Molecular Weight (g/mol) | 203.24 |
| MDL Number | MFCD00037267 |
| InChI Key | PNFVIPIQXAIUAY-UHFFFAOYNA-N |
| Synonym | boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid |
| PubChem CID | 2755934 |
| IUPAC Name | 2-{[(tert-butoxy)carbonyl]amino}butanoic acid |
| SMILES | CCC(NC(=O)OC(C)(C)C)C(O)=O |
Chemical Identifiers
| 34306-42-8 | |
| 203.24 | |
| PNFVIPIQXAIUAY-UHFFFAOYNA-N | |
| 2755934 | |
| CCC(NC(=O)OC(C)(C)C)C(O)=O |
| C9H17NO4 | |
| MFCD00037267 | |
| boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid | |
| 2-{[(tert-butoxy)carbonyl]amino}butanoic acid |
Specifications
| 34306-42-8 | |
| White | |
| MFCD00037267 | |
| boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid | |
| CCC(NC(=O)OC(C)(C)C)C(O)=O | |
| 203.24 | |
| 203.24 | |
| Crystalline Powder |
| 75°C | |
| C9H17NO4 | |
| 5 g | |
| PNFVIPIQXAIUAY-UHFFFAOYNA-N | |
| 2-{[(tert-butoxy)carbonyl]amino}butanoic acid | |
| 2755934 | |
| ≥98.0% (HPLC,T) | |
| (S)-2-(tert-Butoxycarbonylamino)butyric Acid |
Safety and Handling
TSCA : No