missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7123581
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45015G 5 g
B450125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45015G Supplier TCI America Supplier No. B45015G

Chemical Identifiers

CAS 78879-20-6
Molecular Formula C15H18NO4
Molecular Weight (g/mol) 276.31
MDL Number MFCD00143845
InChI Key HFPVZPNLMJDJFB-LBPRGKRZSA-M
Synonym boc-tic-oh, s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-n-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, boc-l-tic-oh, s-2-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, n-boc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-2-methylpropan-2-yl oxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid
PubChem CID 664088
IUPAC Name (3S)-2-[(tert-butoxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@H]1C([O-])=O

Specifications

Melting Point 131°C
Color White
Quantity 5 g
Formula Weight 277.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid
TSCA No
Recommended Storage Refrigerator