missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylpropionic Acid 98.0+%, TCI America™

Catalog No. p-7135016
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
P12201G 1 g
P12205G 5 g
2 options

Catalog No. P12201G

Supplier: TCI America P12201G

Only null left

Chemical Identifiers

CAS 7782-24-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00063139
InChI Key YPGCWEMNNLXISK-ZETCQYMHSA-N
Synonym s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc
PubChem CID 2724622
ChEBI CHEBI:48527
IUPAC Name (2S)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-(+)-2-Phenylpropionic Acid
EINECSNumber (3)-1751
TSCA No