missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylpropionic acid, 99%, Thermo Scientific™

Numéro de catalogue. AC264652500
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC264652500 250mg Glass Bottle
AC264650010 1g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC264652500 Fournisseur Thermo Scientific Chemicals Code fournisseur 264652500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 7782-24-3
Formule moléculaire C9H10O2
Poids moléculaire (g/mol) 150.18
Numéro MDL MFCD00063139
Clé InChI YPGCWEMNNLXISK-ZETCQYMHSA-N
Synonyme s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc
PubChem CID 2724622
ChEBI CHEBI:48527
Nom de l’IUPAC (2S)-2-phenylpropanoic acid
SOURIRES CC(C1=CC=CC=C1)C(=O)O

Spécifications

Nom ou substance chimique (S)-(+)-2-Phenylpropionic acid
Nom Remarque 99%
CAS % minimum 98.5
CAS % maximal 100.0
Densité 1.1000g/mL
Point d’ébullition 115°C (1.0mmHg)
Point d’éclair >110°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98.5% min. (GC)
Emballage Glass Bottle
Indice de réfraction 1.5223
Formule linéaire CH3CH(C6H5)CO2H
Quantité 250mg
Beilstein 09, 347
Gravité spécifique 1.1
Conditions spécifiques de rotation +75° (20°C c=3,CHCl3)
Rotation spécifique + 75
Poids de formule 150.18
Pourcentage de pureté 99%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only