missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylpiperazine 97.0+%, TCI America™

Catalog No. p-7132723
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14241G 1 g
M14245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14241G Supplier TCI America Supplier No. M14241G
Only null left

Chemical Identifiers

CAS 74879-18-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00171452
InChI Key JOMNTHCQHJPVAZ-YFKPBYRVSA-N
Synonym s-+-2-methylpiperazine, s-2-methylpiperazine, 2s-2-methylpiperazine, piperazine, 2-methyl-, 2s, s-2-methyl-piperazine, unii-c5vi3s1yc0, 2-methylpiperazine, s, 2-s-methylpiperazine, s-mp, c5vi3s1yc0
PubChem CID 2734219
IUPAC Name (2S)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 1 g
UN Number 2925
Formula Weight 100.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Methylpiperazine
EINECSNumber (5)-3829
RTECSNumber TM1225020
TSCA No