missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylbutyric acid, 98%

Catalog No. AC224190250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224190250 25 g Glass bottle
AC224190010 1 g Glass bottle
AC224190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC224190250 Supplier Thermo Scientific Chemicals Supplier No. 224190250
Only null left

CAS: 1730-91-2 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1730-91-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00063166
InChI Key WLAMNBDJUVNPJU-BYPYZUCNSA-N
Synonym s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
PubChem CID 448893
ChEBI CHEBI:38655
IUPAC Name (2S)-2-methylbutanoic acid
SMILES CCC(C)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Boiling Point 78.0°C (15.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4041 to 1.4061
Linear Formula CH3CH2CH(CH3)COOH
Quantity 25 g
Beilstein 02, 304
Specific Gravity 0.938
Specific Rotation Condition +19° (20°C c=neat)
Specific Rotation + 19.00
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methylbutyric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only