missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%

Catalog No. AAL0960803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0960803 1 g
AAL0960806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0960803 Supplier Thermo Scientific Chemicals Supplier No. L0960803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63126-47-6 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63126-47-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00010408
InChI Key CHPRFKYDQRKRRK-LURJTMIESA-N
Synonym s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane
PubChem CID 671217
IUPAC Name (2S)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

Density 0.933
Boiling Point 61°C to 62°C (40 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.4457
Quantity 1 g
UN Number UN1993
Beilstein 3587324
Sensitivity Air Sensitive
Optical Rotation +2.2° (c=2 in Benzene)
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 626-212-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only