missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Isopropylamino-3-methyl-1-butanol 97.0+%, TCI America™

Catalog No. I04805G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04805G Supplier TCI America Supplier No. I04805G

Chemical Identifiers

CAS 112211-88-8
Molecular Formula C8H19NO
Molecular Weight (g/mol) 145.246
MDL Number MFCD00671550
InChI Key ANZBKMZVBJDTEL-MRVPVSSYSA-N
Synonym (S)-N-Isopropyl-2-(3-methyl-1-butanol)amine
PubChem CID 13778611
IUPAC Name (2S)-3-methyl-2-(propan-2-ylamino)butan-1-ol
SMILES CC(C)C(CO)NC(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 145.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-2-Isopropylamino-3-methyl-1-butanol