missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Hydroxy-4-phthalimidobutyric Acid 98.0+%, TCI America™

Catalog No. H09335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09335G Supplier TCI America Supplier No. H09335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 48172-10-7
Molecular Formula C12H10NO5
Molecular Weight (g/mol) 248.22
MDL Number MFCD00192213
InChI Key YWDXODQRCDEZLN-VIFPVBQESA-M
Synonym s-+-2-hydroxy-4-phthalimidobutyric acid, s-4-1,3-dioxoisoindolin-2-yl-2-hydroxybutanoic acid, 2s-4-1,3-dioxoisoindolin-2-yl-2-hydroxybutanoic acid, s-+-alpha-hydroxy-1,3-dioxo-2-isoindolinebutyric acid, 2s-4-1,3-dioxoisoindol-2-yl-2-hydroxybutanoic acid, pubchem6049, s-+-a-hydroxy-1,3-dioxo-2-isoindolinebutyric acid, s-4-phthalimido-2-hydroxybutyric acid, s-2-hydroxy-4-phthalimidobutyric acid
PubChem CID 688218
IUPAC Name (2S)-4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-2-hydroxybutanoate
SMILES O[C@@H](CCN1C(=O)C2=CC=CC=C2C1=O)C([O-])=O

Specifications

Melting Point 152°C
Color White
Quantity 5 g
Formula Weight 249.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Hydroxy-4-phthalimidobutyric Acid