missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Hydroxy-3-buten-1-yl p-tosylate, 99%, Thermo Scientific™
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 1g |
| Specific Rotation Condition | − 78.50 (20.00°C c=1.13 MeOH) |
| Specific Rotation | − 78.50 |
| Formula Weight | 242.3 |
| Percent Purity | 99% |
| Chemical Name or Material | (S)-2-Hydroxy-3-buten-1-yl p-tosylate |
RUO – Research Use Only