missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxy-3-buten-1-yl p-tosylate, 99%, Thermo Scientific™

Catalog No. p-4509604
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411970010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC411970010 Supplier Thermo Scientific Chemicals Supplier No. 411970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133095-74-6
Molecular Formula C11H14O4S
Molecular Weight (g/mol) 242.29
MDL Number MFCD00145259
InChI Key YQSCDBZHHLIPOI-JTQLQIEISA-N
Synonym s-3-butene-1,2-diol-1-p-toluenesulfonate, s-2-hydroxy-3-butenyl tosylate, 3-butene-1,2-diol, 1-4-methylbenzenesulfonate , 2s, 2s-1-4-methylbenzenesulfonyl oxy but-3-en-2-ol, pubchem6792, s-3-butene-1,2-diol 1-tosylate, 2s-2-hydroxybut-3-enyl 4-methylbenzenesulfonate, s-4-4-methylphenyl sulfonyl oxy-1-butene-3-ol, s-toluene-4-sulfonic acid 2-hydroxy-but-3-enyl ester, 3-butene-1,2-diol,1-4-methylbenzenesulfonate , 2s
PubChem CID 7015761
IUPAC Name [(2S)-2-hydroxybut-3-enyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H](O)C=C

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1g
Specific Rotation Condition − 78.50 (20.00°C c=1.13 MeOH)
Specific Rotation − 78.50
Formula Weight 242.3
Percent Purity 99%
Chemical Name or Material (S)-2-Hydroxy-3-buten-1-yl p-tosylate

RUO – Research Use Only