missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%

Catalog No. p-7044909
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08692MD 250 mg
AAL0869203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08692MD Supplier Thermo Scientific Chemicals Supplier No. L08692MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13113-71-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13113-71-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00067700
InChI Key NWCHELUCVWSRRS-UHFFFAOYNA-N
Synonym s-atrolactic acid, 2s-2-hydroxy-2-phenylpropanoic acid, s-+-2-hydroxy-2-phenylpropionic acid, atrolactic acid, +, s-2-hydroxy-2-phenylpropanoic acid, unii-22y6g519ru, s-+-atrolactic acid, atrolactic acid 2-phenyl-lactic acid, benzeneacetic acid, a-hydroxy-a-methyl-, as
PubChem CID 445144
ChEBI CHEBI:40741
IUPAC Name (2S)-2-hydroxy-2-phenylpropanoic acid
SMILES CC(O)(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 115°C to 117°C
Quantity 250 mg
Beilstein 2208529
Optical Rotation +49° (c=2 in Water)
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Hydroxy-2-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only