missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Hexanol, 98%

Catalog No. p-7045354
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL10401MD 250 mg
AAL1040103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL10401MD Supplier Thermo Scientific Chemicals Supplier No. L10401MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52019-78-0 | C6H14O | 102.18 g/mol

S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 52019-78-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00065955
InChI Key QNVRIHYSUZMSGM-UHFFFAOYNA-N
Synonym s-+-2-hexanol, s-2-hexanol, 2s-hexan-2-ol, 2-hexanol #, pubchem6732, 2s-2-hexanol, s-hexan-2-ol, s +-2-hexanol
PubChem CID 638097
IUPAC Name (2S)-hexan-2-ol
SMILES CCCCC(C)O

Specifications

Density 0.818
Boiling Point 137°C to 138°C
Flash Point 45°C (113°F)
Refractive Index 1.414
Quantity 250 mg
UN Number UN2282
Beilstein 1718997
Solubility Information Slightly soluble in water.
Optical Rotation +10.5° (Neat)
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Hexanol
Show More Show Less
Hazard Category H226-H302-H319
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only