missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic Acid 98.0+%, TCI America™

Catalog No. C13735G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13735G 5 g
C13731G 1 g
C137325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C13735G Supplier TCI America Supplier No. C13735G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoate
SMILES C[C@H](Cl)C([O-])=O

Specifications

Color Yellow
Quantity 5 g
UN Number 2511
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Chloropropionic Acid
EINECSNumber (2)-1157
RTECSNumber UA2451950
TSCA No