missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic acid, 98%

Catalog No. aal0942714
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0942714 25 g
AAL0942706 5 g
2 options

Catalog No. AAL0942714

Supplier: Thermo Scientific Chemicals L0942714

Only null left

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol

Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

Solubility
Fully miscible in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O

Specifications

Melting Point 4°C
Density 1.27
Boiling Point 187°C
Flash Point 107°C (225°F)
Refractive Index 1.4345
Quantity 25 g
UN Number UN2511
Beilstein 1720257
Solubility Information Fully miscible in water.
Formula Weight 108.53
Percent Purity 99%
Chemical Name or Material (S)-(-)-2-Chloropropionic acid
Show More Show Less
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: 2-CHLOROPROPIONIC ACID
EINECSNumber 411-150-5
RTECSNumber UA2451950
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only