missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic acid, 98%

Catalog No. AC254970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC254970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC254970050 Supplier Thermo Scientific Chemicals Supplier No. 254970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4°C
Color Yellow
Density 1.2400g/mL
Boiling Point 187°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.436
Quantity 5 g
Specific Gravity 1.24
Specific Rotation -14.5
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 108.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Chloropropionic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber UA2451950
TSCA TSCA

RUO – Research Use Only