missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Chloro-3-methylbutyric Acid 98.0+%, TCI America™

Catalog No. C13651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13651G 1 g
C13655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13651G Supplier TCI America Supplier No. C13651G

Chemical Identifiers

CAS 26782-74-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00067109
InChI Key DDTJFSPKEIAZAM-BYPYZUCNSA-N
Synonym (S)-2-Chloroisovaleric Acid
PubChem CID 5324925
IUPAC Name (2S)-2-chloro-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 109°C
Quantity 1 g
UN Number 3265
Formula Weight 136.58
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-2-Chloro-3-methylbutyric Acid