missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Butanol 98.0+%, TCI America™

Catalog No. B09251ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09251ML 1 mL
B09255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09251ML Supplier TCI America Supplier No. B09251ML

Chemical Identifiers

CAS 4221-99-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064281
InChI Key BTANRVKWQNVYAZ-BYPYZUCNSA-N
Synonym s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
PubChem CID 444683
ChEBI CHEBI:45475
IUPAC Name (2S)-butan-2-ol
SMILES CCC(C)O

Specifications

Color Colorless
Quantity 1 mL
Physical Form Clear Liquid at 20°C
UN Number 1120
Boiling Point 100°C
Formula Weight 74.12
Percent Purity ≥98.0% (GC)
Chemical Name or Material (S)-(+)-2-Butanol
EINECSNumber (2)-3049
RTECSNumber EO1750000
TSCA No