missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)butyric acid, 95%

Catalog No. AAH6368614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6368614 25 g
AAH6368603 1 g
AAH6368606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6368614 Supplier Thermo Scientific Chemicals Supplier No. H6368614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34306-42-8 | C9H17NO4 | 203.24 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 34306-42-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037267
InChI Key PNFVIPIQXAIUAY-UHFFFAOYNA-N
Synonym boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
PubChem CID 2755934
SMILES CCC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 25 g
Beilstein 6801706
Solubility Information Slightly soluble in water.
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Formula Weight 203.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (S)-2-(Boc-amino)butyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only