missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%

Catalog No. p-7034295
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196603 1 g
AAH5196606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196603 Supplier Thermo Scientific Chemicals Supplier No. H5196603

CAS: 100564-78-1 | C15H21NO4 | 279.336 g/mol

(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 100564-78-1
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00076904
InChI Key MCODLPJUFHPVQP-LBPRGKRZSA-N
Synonym boc-l-homophenylalanine, boc-homophe-oh, boc-hophe-oh, boc-l-homo-phe, s-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid, boc-homophenylalanine, boc-hfe-oh, n-boc-l-homophenylalanine, n-alpha-boc-l-homophenylalanine
PubChem CID 7018726
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 76°C to 80°C
Beilstein 3653506
Solubility Information Soluble in methanol.
Quantity 1 g
Optical Rotation +6.5° (c=2 in Ethanol)
Formula Weight 279.34
Percent Purity 98%
Chemical Name or Material (S)-2-(Boc-amino)-4-phenylbutyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only