missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%

Catalog No. p-7054512
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6277203 1 g
AAH6277206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6277203 Supplier Thermo Scientific Chemicals Supplier No. H6277203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3350-13-8 | C29H47N3O6 | 533.71 g/mol

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 3350-13-8
Molecular Formula C29H47N3O6
Molecular Weight (g/mol) 533.71
MDL Number MFCD00237341
InChI Key CYMIEBREXUYHGN-UHFFFAOYNA-N
Synonym dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha
PubChem CID 49853427
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 553.7
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only