missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Benzyl-1-Boc-piperazine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5278503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5278503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5278503 Supplier Thermo Scientific Chemicals Supplier No. H5278503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 169447-86-3 | Molecular Formula: C16H24N2O2 | Molecular Weight: 276.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 169447-86-3
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD03787923
InChI Key QKUHUJCLUFLGCI-AWEZNQCLSA-N
Synonym s-1-boc-2-benzylpiperazine, s-1-boc-2-benzyl-piperazine, s-tert-butyl 2-benzylpiperazine-1-carboxylate, tert-butyl 2s-2-benzylpiperazine-1-carboxylate, s-n1-boc-2-benzylpiperazine, 2 s-benzyl-piperazine-1-carboxylic acid tert-butyl ester, s-2-benzyl-piperazine-1-carboxylic acid tert-butyl ester, n1-boc-2-benzylpiperazine, s-1-n-boc-2-benzylpiperazine, s-2-benzyl-1-boc-piperazine
PubChem CID 17750441
IUPAC Name tert-butyl (2S)-2-benzylpiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC[C@@H]1CC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 276.38
Percent Purity 97%
Chemical Name or Material (S)-2-Benzyl-1-Boc-piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only