missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, Thermo Scientific Chemicals

Catalog No. AC105142500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105142500 250 mg Glass bottle
AC105140010 1 g Glass bottle
AC105140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105142500 Supplier Thermo Scientific Chemicals Supplier No. 105142500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2133-34-8 | C4H7NO2 | 101.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2133-34-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
InChI Key IADUEWIQBXOCDZ-VKHMYHEASA-N
Synonym l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
PubChem CID 16486
ChEBI CHEBI:6198
IUPAC Name (2S)-azetidine-2-carboxylic acid
SMILES C1CNC1C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 206°C to 207°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Merck Index 10,911
Specific Rotation -125
Solubility Information Solubility in water: 5% in water
Formula Weight 101.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
Show More Show Less
EINECSNumber 218-362-5

RUO – Research Use Only