missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Anilinomethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. A09451G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A09451G 1 g
A09455G 5 g
2 options

Catalog No. A09451G

Supplier: TCI America A09451G

Only null left

Chemical Identifiers

CAS 64030-44-0
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00015890
InChI Key MCHWKJRTMPIHRA-NSHDSACASA-N
Synonym s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
PubChem CID 6950385
IUPAC Name N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
SMILES C1CC(NC1)CNC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 126°C
Quantity 1 g
UN Number 1760
Formula Weight 176.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-(Anilinomethyl)pyrrolidine