missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminononane, ChiPros 99+%, ee 99+%

CAS: 869278-88-6 | C9H21N | 143.27 g/mol

$96.21 - $1105.08

Chemical Identifiers

CAS 869278-88-6
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD03844738
InChI Key ALXIFCUEJWCQQL-UHFFFAOYNA-N
Synonym 2-nonanamine, 2-aminononane, 2-nonylamine, 2-amino-nonane, acmc-20apfg, acmc-1bh6g
PubChem CID 12878166
IUPAC Name nonan-2-amine
SMILES CCCCCCCC(C)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1931103
View Documents
Thermo Scientific Chemicals
L1931103
1 g
Each for $96.21
Only null left
 
AAL1931106
View Documents
Thermo Scientific Chemicals
L1931106
5 g
Each for $285.99
Only null left
 
AAL1931114
View Documents
Thermo Scientific Chemicals
L1931114
25 g
Each for $1,105.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

869278-88-6
143.27
ALXIFCUEJWCQQL-UHFFFAOYNA-N
12878166
CCCCCCCC(C)N
C9H21N
MFCD03844738
2-nonanamine, 2-aminononane, 2-nonylamine, 2-amino-nonane, acmc-20apfg, acmc-1bh6g
nonan-2-amine

Specifications

869278-88-6
73°C (19 mmHg)
C9H21N
MFCD03844738
UN2735
2-nonanamine, 2-aminononane, 2-nonylamine, 2-amino-nonane, acmc-20apfg, acmc-1bh6g
+6° (Neat)
nonan-2-amine
12878166
≥99%
ee ≥99%
0.782
71°C (159°F)
1.4271
1 g
Air Sensitive
ALXIFCUEJWCQQL-UHFFFAOYNA-N
CCCCCCCC(C)N
143.27
143.27
(S)-(+)-2-Aminononane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 663-044-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only