missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine, 95%

Catalog No. AAH5713703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5713703 1 g
AAH5713706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5713703 Supplier Thermo Scientific Chemicals Supplier No. H5713703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22795-99-9 | C7H16N2 | 128.219 g/mol

It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Hygroscopic. Store away from acids, water/moisture, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

Chemical Identifiers

CAS 22795-99-9
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00191371
InChI Key UNRBEYYLYRXYCG-ZETCQYMHSA-N
Synonym s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590
PubChem CID 643457
IUPAC Name [(2S)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

Density 0.919
Boiling Point 50°C to 54°C
Flash Point 57°C (135°F)
Refractive Index 1.466
Quantity 1 g
UN Number UN2733
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform,methanol. Slightly soluble in water.
Optical Rotation −95° (c=1 in Methanol)
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
Show More Show Less
Hazard Category H226-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only