missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid, 98%, 94% ee

Catalog No. p-4510091
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299142500 250 mg Glass bottle
AC299140010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC299142500 Supplier Thermo Scientific Chemicals Supplier No. 299142500

CAS: 2280-27-5 | C5H11NO3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2280-27-5
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
InChI Key LDRFQSZFVGJGGP-GSVOUGTGSA-N
Synonym s-2-amino-3-hydroxy-3-methylbutanoic acid, 3-hydroxy-l-valine, s-+-2-amino-3-hydroxy-3-methylbutanoic acid, beta-hydroxy-l-valine, valine, 3-hydroxy, 2s-2-amino-3-hydroxy-3-methylbutanoic acid, 2s-2-amino-3-hydroxy-3-methyl-butanoic acid, 3-hydroxyvaline, 3-methylthreonine, 3-hydroxy-l-val
PubChem CID 192763
IUPAC Name (2S)-2-amino-3-hydroxy-3-methylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)O

Specifications

Color White
Enantiomeric Excess 93% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3C(CH3)(OH)CH(NH2)COOH
Quantity 250 mg
Specific Rotation Condition − 11.75 (25.00°C c=2,6N HCl)
Specific Rotation − 11.75
Formula Weight 133.15
Percent Purity 98%,94% ee
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid
Show More Show Less

RUO – Research Use Only