missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-butanol 98.0+%, TCI America™

Catalog No. A09735ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09735ML 5 mL
A09731ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09735ML Supplier TCI America Supplier No. A09735ML

Chemical Identifiers

CAS 5856-62-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00064418
InChI Key JCBPETKZIGVZRE-BYPYZUCNSA-N
Synonym s-+-2-amino-1-butanol, s-2-aminobutan-1-ol, 2s-2-aminobutan-1-ol, s-2-aminobutanol, unii-98jem4yebr, s-2-amino-1-butanol, +-2-amino-1-butanol, s +-2-amino-1-butanol, 2-amino-1-butanol, +
PubChem CID 120084
IUPAC Name (2S)-2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Color Yellow
Boiling Point 178°C
Quantity 5 mL
UN Number 3267
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Amino-1-butanol
EINECSNumber (2)-0316
TSCA No