missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-butanol, 98+%

CAS: 5856-62-2 | C4H11NO | 89.138 g/mol

$44.14 - $147.04

Chemical Identifiers

CAS 5856-62-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00064418
InChI Key JCBPETKZIGVZRE-BYPYZUCNSA-N
Synonym s-+-2-amino-1-butanol, s-2-aminobutan-1-ol, 2s-2-aminobutan-1-ol, s-2-aminobutanol, unii-98jem4yebr, s-2-amino-1-butanol, +-2-amino-1-butanol, s +-2-amino-1-butanol, 2-amino-1-butanol, +
PubChem CID 120084
IUPAC Name (2S)-2-aminobutan-1-ol
SMILES CCC(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1144903
View Documents
Thermo Scientific Chemicals
L1144903
1 g
Each for $44.14
Only null left
 
AAL1144906
View Documents
Thermo Scientific Chemicals
L1144906
5 g
Each for $147.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5856-62-2
89.138
JCBPETKZIGVZRE-BYPYZUCNSA-N
120084
CCC(CO)N
C4H11NO
MFCD00064418
s-+-2-amino-1-butanol, s-2-aminobutan-1-ol, 2s-2-aminobutan-1-ol, s-2-aminobutanol, unii-98jem4yebr, s-2-amino-1-butanol, +-2-amino-1-butanol, s +-2-amino-1-butanol, 2-amino-1-butanol, +
(2S)-2-aminobutan-1-ol

Specifications

5856-62-2
11.1
89°C (192°F)
C4H11NO
MFCD00064418
UN2735
Air sensitive; Hygroscopic
JCBPETKZIGVZRE-BYPYZUCNSA-N
CCC(CO)N
89.138
89.14
(S)-(+)-2-Amino-1-butanol
0.947
176°C to 178°C
Amine-like
1.4525
1 g
1718930
s-+-2-amino-1-butanol, s-2-aminobutan-1-ol, 2s-2-aminobutan-1-ol, s-2-aminobutanol, unii-98jem4yebr, s-2-amino-1-butanol, +-2-amino-1-butanol, s +-2-amino-1-butanol, 2-amino-1-butanol, +
+12° (c=2 in Ethanol)
(2S)-2-aminobutan-1-ol
120084
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 227-475-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only