missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-butanol, 98+%

Catalog No. AAL1144903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1144903 1 g
AAL1144906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1144903 Supplier Thermo Scientific Chemicals Supplier No. L1144903
Only null left

CAS: 5856-62-2 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5856-62-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00064418
InChI Key JCBPETKZIGVZRE-BYPYZUCNSA-N
Synonym s-+-2-amino-1-butanol, s-2-aminobutan-1-ol, 2s-2-aminobutan-1-ol, s-2-aminobutanol, unii-98jem4yebr, s-2-amino-1-butanol, +-2-amino-1-butanol, s +-2-amino-1-butanol, 2-amino-1-butanol, +
PubChem CID 120084
IUPAC Name (2S)-2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Density 0.947
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 1 g
UN Number UN2735
Beilstein 1718930
Sensitivity Air sensitive; Hygroscopic
Optical Rotation +12° (c=2 in Ethanol)
Formula Weight 89.14
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Amino-1-butanol
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 227-475-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only