missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic Acid 99.0+%, TCI America™

Catalog No. M102125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M102125G 25 g
M1021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M102125G Supplier TCI America Supplier No. M102125G

Chemical Identifiers

CAS 22204-53-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD00010500
InChI Key CMWTZPSULFXXJA-VIFPVBQESA-N
Synonym naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno
PubChem CID 156391
ChEBI CHEBI:7476
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 230.26
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic Acid
RTECSNumber UF5275000
TSCA No