missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,4-Diphenyl-4,5-dihydrooxazole 98.0+%, TCI America™

$269.52 - $816.28

Chemical Identifiers

CAS 475571-19-8
Molecular Formula C15H13NO
Synonym (S)-Pheox
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D53681G
View Documents
TCI America
D53681G
1 g
Each for $269.52
Only null left
 
D53685G
View Documents
TCI America
D53685G
5 g
Each for $816.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

475571-19-8
C15H13NO
(S)-Pheox

Specifications

475571-19-8
C15H13NO
(S)-Pheox
≥98.0% (GC)
(S)-2,4-Diphenyl-4,5-dihydrooxazole
White-Yellow
1 g
223.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'