missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2,4-Diphenyl-4,5-dihydrooxazole 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 223.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (S)-2,4-Diphenyl-4,5-dihydrooxazole |
Safety and Handling
| TSCA | No |