missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,3-O-Isopropylideneglycerol, 98%

Catalog No. p-7027603
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2448804 2 g
AAB2448809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2448804 Supplier Thermo Scientific Chemicals Supplier No. B2448804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22323-82-6 | C6H12O3 | 132.16 g/mol

(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.

Chemical Identifiers

CAS 22323-82-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00063239
InChI Key RNVYQYLELCKWAN-UHFFFAOYNA-N
Synonym s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol
PubChem CID 736057
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(C)OCC(CO)O1

Specifications

Density 1.066
Boiling Point 82°C to 83°C (14 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.435
Quantity 2 g
Beilstein 80118
Merck Index 14,5213
Solubility Information Miscible with water and chloroform.
Optical Rotation +11.5° (c=5 in Methanol)
Formula Weight 132.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2,3-O-Isopropylideneglycerol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 244-910-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only