Learn More
(S)-2,3-Diaminopropionic acid monohydrochloride, 95%
CAS: 1482-97-9 | C3H9ClN2O2 | 140.567 g/mol
Supplier: Thermo Scientific Chemicals H6212806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-2,3-Diaminopropionic acid monohydrochloride | |
| 231°C to 233°C (decomposition) | |
| C3H9ClN2O2 | |
| 5 g | |
| Hygroscopic | |
| SKWCZPYWFRTSDD-DKWTVANSSA-N | |
| (2S)-2,3-diaminopropanoic acid;hydrochloride | |
| 12495315 | |
| 95% |
| 1482-97-9 | |
| White | |
| MFCD00065497 | |
| 4263944 | |
| 3-amino-l-alanine hydrochloride, s-2,3-diaminopropanoic acid hydrochloride, h-dap-oh.hcl, l-2,3-diaminopropionic acid hydrochloride, l +-2,3-diaminopropionic acid hydrochloride, l-3-aminoalanine hydrochloride, h-dapa-oh.hcl, s-+-2,3-diaminopropionic acid hydrochloride, l-alanine, 3-amino-, monohydrochloride, 2,3-diaminopropionic acid hydrochloride, + | |
| C(C(C(=O)O)N)N.Cl | |
| 140.567 | |
| 140.57 | |
| Crystalline Solid |
Chemical Identifiers
| 1482-97-9 | |
| 140.567 | |
| SKWCZPYWFRTSDD-DKWTVANSSA-N | |
| 12495315 | |
| C(C(C(=O)O)N)N.Cl |
| C3H9ClN2O2 | |
| MFCD00065497 | |
| 3-amino-l-alanine hydrochloride, s-2,3-diaminopropanoic acid hydrochloride, h-dap-oh.hcl, l-2,3-diaminopropionic acid hydrochloride, l +-2,3-diaminopropionic acid hydrochloride, l-3-aminoalanine hydrochloride, h-dapa-oh.hcl, s-+-2,3-diaminopropionic acid hydrochloride, l-alanine, 3-amino-, monohydrochloride, 2,3-diaminopropionic acid hydrochloride, + | |
| (2S)-2,3-diaminopropanoic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only