missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,3-Diaminopropionic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7126699
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1377100MG 100 mg
D13771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1377100MG Supplier TCI America Supplier No. D1377100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1482-97-9
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00065497
InChI Key SKWCZPYWFRTSDD-DKWTVANSSA-N
Synonym 3-amino-l-alanine hydrochloride, s-2,3-diaminopropanoic acid hydrochloride, h-dap-oh.hcl, l-2,3-diaminopropionic acid hydrochloride, l +-2,3-diaminopropionic acid hydrochloride, l-3-aminoalanine hydrochloride, h-dapa-oh.hcl, s-+-2,3-diaminopropionic acid hydrochloride, l-alanine, 3-amino-, monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +
PubChem CID 12495315
IUPAC Name (2S)-2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 100 mg
Formula Weight 140.57
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2,3-Diaminopropionic Acid Hydrochloride