missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid 98.0+%, TCI America™
$117.43 - $380.27
Chemical Identifiers
| CAS | 14590-53-5 |
|---|---|
| Molecular Formula | C6H10O2 |
| Molecular Weight (g/mol) | 114.144 |
| MDL Number | MFCD08460918 |
| InChI Key | BFNMOMYTTGHNGJ-SCSAIBSYSA-N |
| PubChem CID | 10192561 |
| IUPAC Name | (1S)-2,2-dimethylcyclopropane-1-carboxylic acid |
| SMILES | CC1(CC1C(=O)O)C |
Chemical Identifiers
| 14590-53-5 | |
| 114.144 | |
| BFNMOMYTTGHNGJ-SCSAIBSYSA-N | |
| (1S)-2,2-dimethylcyclopropane-1-carboxylic acid |
| C6H10O2 | |
| MFCD08460918 | |
| 10192561 | |
| CC1(CC1C(=O)O)C |
Specifications
| 14590-53-5 | |
| 100°C | |
| MFCD08460918 | |
| BFNMOMYTTGHNGJ-SCSAIBSYSA-N | |
| (1S)-2,2-dimethylcyclopropane-1-carboxylic acid | |
| 10192561 | |
| ≥98.0% (GC,T) | |
| (S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid |
| Yellow | |
| C6H10O2 | |
| 1 g | |
| CC1(CC1C(=O)O)C | |
| 114.144 | |
| 114.14 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-3774
TSCA : No